Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium iodate reagent g500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 207977500G
Encompass
KIO3 can be used as a substitute of KI in radiation protection. A kinetic study of thermal degradation of KIO3 by³-rays suggests that rate of decomposition increases while activation energy decreases upon irradiation.
Catalog No. 50-289-8933