Learn More
Alkali Scientific Potassium Iodide, 500 Grams
SDP
Supplier:  Alkali Scientific UMI002CP
Encompass_Preferred
Potassium Iodide, laboratory grade, is a chemical reagent available in a 500g quantity, commonly used in analytical chemistry and biochemistry. It serves as a key component in a variety of applications, including the preparation of iodine solutions, titrations, and the formation of iodine complexes. Potassium iodide is also employed in research on redox reactions, where it is used as a reducing agent. Additionally, it plays a role in the synthesis of organic iodides and in the iodination of compounds for pharmaceutical applications. This reagent is widely used in chemical analysis, medical research, and laboratory-based applications that require iodine compounds.
Catalog No. 50-006-61733