Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium iodide ACS reage12KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 22194512KG
Encompass
Potassium iodide is a water soluble inorganic salt that can be prepared by reacting hydrogen iodide and potassium bicarbonate.
Catalog No. 50-290-3673