Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium iodide ReagentPl500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 207969500G
Encompass
Potassium iodide (KI) is an inorganic potassium salt. KI is a colorless solid having cubic crystal system. It has been prepared by reacting hydroiodic acid with potassium hydroxide or potassium carbonate. Alumina loaded with potassium iodide has been employed as catalyst for the preparation of biodiesel by the transesterification reaction of soybean oil with methanol.
Catalog No. 50-289-7795