Learn More
Medchemexpress LLC Potassium oxonate | 2207-75-2 | 195.17 | 10MM/1ML
SDP
Supplier:  Medchemexpress LLC HY1751110MM/1ML
Encompass_Preferred
Potassium oxonate, also known as Potassium azaorotate, is a selective competitive uricase inhibitor. It functions by inhibiting the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate, blocking the action of hepatic uricase.
  • Inhibits phosphorylation of 5-FU
  • Blocks action of hepatic uricase
  • Used in animal modeling to create hyperuricemia models
  • Targets xanthine oxidase
  • Relevant to metabolic disease research
Catalog No. 50-003-86282