Learn More
Medchemexpress LLC Potassium oxonate | 2207-75-2 | 195.17 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY17511
Encompass_Preferred
Potassium oxonate (Potassium azaorotate) is a uricase inhibitor that inhibits the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate. It can also be used in animal modeling to create hyperuricemia models and primarily distributes within the cells of the small intestine after metabolism.
  • Selective competitive uricase inhibitor
  • Blocks the action of hepatic uricase
  • Can be used in animal modeling to create hyperuricemia models
Catalog No. 50-003-85316