Learn More
Medchemexpress LLC Potassium p-nitrophenyl sulfate | 6217-68-1 | 99.89% | 257.26 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW009454500MG
Encompass_Preferred
Potassium p-nitrophenyl sulfate is a chromogenic substrate used for arylsulfatase. It is released by arylsulfonase cleavage, allowing the activity of arylsulfonase to be quantitatively determined by colorimetric detection at 400 nm.
  • Chromogenic substrate for arylsulfatase
  • Supports quantitative determination of arylsulfonase activity
  • Solid appearance
  • Off-white to yellow color
  • Soluble in H2O at 25 mg/mL
Catalog No. 50-004-52096