Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium permanganate ACS25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3991242.5KG
Encompass
Potassium permanganate (PP KMnO4) is a dark violet colored crystalline solid. It is a strong oxidizing agent used in the oxidation of a wide range of organic compounds. PP is commonly used as a disinfectant to treat external infections in aquaculture. Corncobs pretreated with alkaline potassium permanganate SOLUTIONon showed high carbohydrate recovery due to the increased enzymatic hydrolysis of lignocellulose. The acidic potassium permanganate chemiluminescence detection has been employed for the detection of cannabinoids.
Catalog No. 50-289-7878