Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium permanganate ACS500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 223468500G
Encompass
Potassium permanganate a dark violet colored crystalline solid is a strong oxidizing agent for a wide range of organic compounds. It is commonly used as a disinfectant to treat external infections in aquaculture. Corncobs pretreated with alkaline potassium permanganate SOLUTIONon showed high carbohydrate recovery due to the increased enzymatic hydrolysis of lignocellulose. The acidic potassium permanganate acts a chemiluminescence reagent for the determination of organic compounds.
Catalog No. 50-289-7098