Learn More
Strem, An Ascensus Company CAS# 10466-65-6. 2g. Potassium perrhenate (99%-Re). MFCD00011365
SDP
Supplier:  Strem, An Ascensus Company 7505002G
Encompass
CAS# 10466-65-6. 2g. Potassium perrhenate (99%-Re). MFCD00011365. Molecular Weight: 289.30. Molecular Formula: KReO4. Color/form: white xtl. Strem# 75-0500. http://www.strem.com/catalog/v/75-0500/
Catalog No. 50-901-13046