Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 1 Kg
SDP
Supplier:  Alkali Scientific POA010CP
Encompass
Potassium Phosphate Dibasic, Anhydrous, is a laboratory-grade salt used for a variety of biological and chemical applications. This 1 kg package offers a reliable source of potassium and phosphate ions, often used in buffer systems, chromatography, and as a reagent for pH control. The anhydrous form ensures the absence of water, which makes it suitable for precise formulation of buffers and other solutions. It is commonly used in biological assays, including enzyme studies and microbial growth media, as it provides essential ions for cellular processes. Potassium Phosphate Dibasic also plays a role in nutrient solutions, serving as a source of phosphate for plant and cell culture studies.
Catalog No. 50-006-60095