Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 12 Kg
SDP
Supplier:  Alkali Scientific POA012CP
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (12 kg), is a high-purity salt that provides an essential source of potassium and phosphate ions for laboratory applications. The larger 12 kg volume is ideal for research environments that require substantial quantities of this reagent. It is frequently used in the preparation of phosphate buffers for various chemical and biological applications, including enzyme assays, cell culture media, and chromatography. As an anhydrous salt, it offers excellent control over concentration and formulation, ensuring consistency and reproducibility in experiments. This reagent is indispensable in studies of plant growth, microbial metabolism, and biochemical reactions where phosphate plays a critical role.
Catalog No. 50-006-61241