Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 2.5 Kg
SDP
Supplier:  Alkali Scientific POA013CP
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (2.5 kg), is a laboratory-grade reagent used to provide a stable source of phosphate ions in various research applications. This salt is typically employed in the preparation of phosphate buffers, nutrient media, and for controlling the pH of chemical reactions. It is essential in biochemical studies, especially those involving enzymatic reactions, cell culture, and chromatography. The 2.5 kg package is perfect for medium to large-scale applications, providing sufficient quantities for frequent use in laboratory settings. The anhydrous form allows for precise formulation of solutions, ensuring optimal performance in scientific experiments requiring accurate ionic balance.
Catalog No. 50-006-61404