Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 3 Kg
SDP
Supplier:  Alkali Scientific POA015CP
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (3 kg), is a laboratory-grade salt used to prepare phosphate buffer solutions, nutrient media, and for controlling pH in chemical reactions. The 3 kg size is well-suited for medium-volume applications, providing a reliable supply of potassium and phosphate ions for various biochemical assays, microbiological culture media, and plant research. Its anhydrous nature ensures that no water content affects the concentration of solutions, making it an excellent choice for precise buffer preparation. Potassium Phosphate Dibasic is frequently used in studies of enzyme activity, plant growth, and other applications where phosphate is a key component.
Catalog No. 50-006-60131