Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, 5 Kg
SDP
Supplier:  Alkali Scientific POA017CP
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous (5 kg), is a laboratory-grade reagent essential for preparing phosphate buffers, controlling pH, and providing phosphate ions in biochemical and biological research. This salt is commonly used in the preparation of enzyme reaction mixtures, plant culture media, and microbial growth solutions. The 5 kg size provides an ample supply for medium-to-large laboratories, making it ideal for routine use in educational and research environments. As an anhydrous salt, it allows for precise solution formulation without worrying about the moisture content that may vary in hydrated forms. Potassium Phosphate Dibasic is widely used in fields such as microbiology, plant biology, and biochemistry.
Catalog No. 50-006-61229