Learn More
Alkali Scientific Potassium Phosphate, Dibasic, Anhydrous, ACS, 500 G
SDP
Supplier:  Alkali Scientific POA019CP
Encompass_Preferred
Potassium Phosphate Dibasic, Anhydrous, ACS (500 g), is a laboratory-grade reagent that is highly useful for preparing phosphate buffers, pH-controlled solutions, and media for biological studies. The ACS grade ensures superior purity, making it suitable for precise research applications in both academic and commercial laboratories. This 500 g package is ideal for smaller-scale experiments where a reliable source of potassium and phosphate ions is needed. The anhydrous form ensures accurate concentrations of phosphate without the complications of moisture content. Potassium Phosphate Dibasic is commonly used in enzymatic assays, plant studies, and biochemical experiments that require phosphate as a key component.
Catalog No. 50-006-61211