Learn More
Alkali Scientific Potassium Phosphate, Dibasic Trihydrate, 10 Kg
SDP
Supplier:  Alkali Scientific TED004CP
Encompass_Preferred
Potassium Phosphate Dibasic Trihydrate is supplied in a 10Kg package, making it suitable for large-scale laboratory use. This chemical is widely used in the preparation of phosphate buffers, which are essential for maintaining stable pH levels in biological and chemical applications. The tri-hydrate form of potassium phosphate ensures better solubility and stability in aqueous solutions. It is commonly used in protein purification, tissue culture, and other experiments requiring controlled pH. This reagent plays a key role in molecular biology and biochemical research by providing a consistent buffer that supports a wide range of reactions and assays.
Catalog No. 50-006-61284