Learn More
Alkali Scientific Potassium Phosphate, Dibasic Trihydrate, 5 Kg
SDP
Supplier:  Alkali Scientific TED003CP
Encompass_Preferred
Potassium Phosphate Dibasic Trihydrate (K2HPO4·3H2O) is a reagent supplied in a 5Kg pack, commonly used in laboratories for various applications requiring stable phosphate buffering. This reagent is particularly useful for maintaining pH balance in biochemical reactions, as it is effective in a wide pH range. The tri-hydrated form of potassium phosphate is often used in the preparation of buffers, cell culture media, and protein purification. It is also used in the preparation of saline solutions and for other laboratory assays requiring phosphate buffers. Potassium Phosphate Dibasic Trihydrate is an essential chemical in scientific research, pharmaceutical formulations, and clinical diagnostics.
Catalog No. 50-006-60869