Learn More
Alkali Scientific Potassium Phosphate Dibasic Trihydrate, 500 G
SDP
Supplier:  Alkali Scientific TED002CP
Encompass
Potassium Phosphate Dibasic Trihydrate, 500 grams, is a laboratory-grade reagent used for preparing phosphate buffers and pH adjustment in scientific experiments. The hydrated form provides enhanced stability, making it ideal for use in a wide range of biochemical and chemical applications. This reagent is used in the preparation of buffers for enzyme assays, chromatography, and protein analysis. Potassium phosphate dibasic also plays a critical role in maintaining physiological pH in cell culture media and in biological research applications. The 500g package is well-suited for smaller-scale experiments and routine laboratory use, ensuring reliable and consistent performance in buffer preparation and pH control.
Catalog No. 50-006-61251