Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 10 Kg
Supplier: Alkali Scientific POA003CP
Potassium Phosphate Monobasic, ACS Grade, is provided in a 10 kg volume for large-scale applications in laboratory settings. This high-quality salt is primarily used to create phosphate-buffered solutions essential in biological and chemical experiments. Potassium Phosphate Monobasic is commonly used in the preparation of buffer solutions, particularly those needed for pH control in protein assays, nucleic acid extraction, and enzyme catalysis. The 10 kg container is perfect for laboratories with high-demand testing or manufacturing processes that require consistent, reliable buffering performance. The reagent's purity and stable pH properties are crucial for reproducible results in various scientific studies.