Learn More
Hach Company Potassium Phosphate, Monobasic, ACS, 454 g
Supplier:  Hach Company 17001H
Encompass
ACS Reagent grade KH2PO4, CAS No. 7778-77-0 For pH buffer and dilution water preparation
Catalog No. 50-199-7384