Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium phosphate monoba25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P06624X2.5KG
Encompass
Potassium phosphate monobasic (monopotassium orthophosphate) is a water soluble inorganic salt. Its standard enthalpy of formation has been reported to be -376.1kcalmol-1. It acts a starting material in the synthesis of potassium metaphosphate.
Catalog No. 50-291-4614