Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 1 Kg
SDP
Supplier:  Alkali Scientific POA002CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, is a reagent widely used in chemical and biological laboratories. The 1 kg container provides a bulk quantity for larger-scale applications such as buffer preparation, cell culture media, and as a component in phosphate-buffered saline (PBS) solutions. Potassium Phosphate Monobasic helps maintain a stable pH environment, which is essential for accurate and consistent results in many experiments, particularly in biochemical and microbiological studies. The reagent's high purity and solubility make it a critical ingredient for applications in molecular biology, protein electrophoresis, and the preparation of standard buffer solutions for various assays.
Catalog No. 50-006-60955