Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 100 G
SDP
Supplier:  Alkali Scientific POA001CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, is a high-quality laboratory reagent supplied in a 100g volume. It is primarily used as a buffer solution in various biological, chemical, and analytical applications. The salt plays a crucial role in maintaining pH balance during reactions or processes requiring acidic conditions. Potassium Phosphate Monobasic is commonly used in preparing phosphate-buffered saline (PBS) and other buffer systems, especially in enzymatic assays and DNA/RNA extraction procedures. Its purity and consistency make it ideal for experiments that demand high-grade reagents. This 100g container is convenient for small-scale laboratory work, offering ease of use and accurate results.
Catalog No. 50-006-61312