Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 12 Kg
SDP
Supplier:  Alkali Scientific POA004CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, is available in a 12 kg volume, making it ideal for high-volume applications in laboratory and industrial settings. This reagent is essential for creating phosphate-buffered saline (PBS) and other buffer systems used in biological research, chemical reactions, and pharmaceutical formulations. The high solubility and pH stability of Potassium Phosphate Monobasic ensure consistent performance, particularly in experiments where precise pH control is required. It is widely used in microbiology, biochemistry, and molecular biology for protein purification, enzyme reactions, and the maintenance of optimal conditions for cell cultures and DNA/RNA extractions.
Catalog No. 50-006-60978