Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 25 Kg
SDP
Supplier:  Alkali Scientific POA007CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, is available in a 25 kg volume, making it ideal for large-scale laboratory and industrial applications. This reagent is primarily used to prepare phosphate-buffered solutions necessary for chemical reactions, biological assays, and pharmaceutical formulations. Potassium Phosphate Monobasic helps to maintain a stable pH environment, crucial for accurate results in various experimental procedures, particularly in microbiology, biochemistry, and molecular biology. The large 25 kg container is suited for high-volume laboratories or manufacturing processes that require a reliable and cost-effective solution for buffer preparation on a large scale.
Catalog No. 50-006-61221