Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 5 Kg
SDP
Supplier:  Alkali Scientific POA009CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, 5 kg volume, is a versatile and essential laboratory reagent used in preparing phosphate-buffered solutions and for maintaining stable pH levels during chemical and biological experiments. The 5 kg quantity is ideal for high-demand laboratories that require larger volumes of reagent for buffer preparation, microbial culture maintenance, and protein assays. Potassium Phosphate Monobasic is commonly used in molecular biology, cell culture, and microbiology for its ability to provide reliable and consistent pH control, essential for the success of experiments and analysis in a variety of research fields.
Catalog No. 50-006-61549