Learn More
Alkali Scientific Potassium Phosphate, Monobasic, ACS Grade, 500 G
SDP
Supplier:  Alkali Scientific POA008CP
Encompass_Preferred
Potassium Phosphate Monobasic, ACS Grade, 500g volume, is a laboratory-grade reagent used for preparing buffer solutions in biochemical and microbiological applications. The reagent helps maintain stable pH levels in solutions used for enzyme assays, DNA/RNA extraction, and protein purification. Potassium Phosphate Monobasic is a critical component of phosphate-buffered saline (PBS), which is widely used in cell culture, immunology, and molecular biology. The 500g size is suitable for routine laboratory work, providing an appropriate quantity of reagent for daily testing and buffer preparation while ensuring consistency in pH control for experiments requiring precise and reproducible results.
Catalog No. 50-006-61222