Learn More
Alkali Scientific Potassium Phosphate Dibasic Trihydrate, 1 Kg
SDP
Supplier:  Alkali Scientific TED001CP
Encompass
Potassium Phosphate Dibasic Trihydrate, 1 kg, is a laboratory-grade reagent that is primarily used as a buffer in scientific research and chemical reactions. This hydrated form of potassium phosphate is essential for preparing buffers that maintain a consistent pH in a variety of laboratory procedures. It is used in biological and chemical research, particularly in experiments requiring stable and precise pH conditions. Potassium phosphate dibasic is also used in the preparation of phosphate buffers for protein analysis, chromatography, and enzyme reactions. The 1 kg packaging provides ample quantity for routine laboratory applications while maintaining the purity required for precise experimental results.
Catalog No. 50-006-60871