Learn More
Sigma Aldrich Fine Chemicals Biosciences Potassium phosphate tribas
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P562910KG
Encompass
Potassium phosphate tribasic exhibits high catalytic property in trans esterification reactions. It has bactericidal activity.
Catalog No. 50-290-2524