Learn More
Medchemexpress LLC Potassium phosphate tribasic monohydrate | 27176-10-9 | MFCD00150410 | 90.0% | 230.28 g/mol | K3PO4.H2O | 1 KG

Supplier: Medchemexpress LLC HYW0897751KG
Potassium phosphate tribasic monohydrate (90%) is an inorganic salt supplied as a monohydrate solid for biochemical assays and laboratory buffer preparation. It is intended for research use only.
- Purity 90.0%.
- Chemical formula K3PO4.H2O.
- Molecular weight 230.28 g/mol.
- CAS number 27176-10-9.
- Available packaging 100 g, 500 g, 1 kg.
- Storage store at room temperature; in solvent store at -80°C for 2 years or -20°C for 1 year.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.