Learn More
Medchemexpress LLC Potassium pyrosulfate | 7790-62-7 | MFCD00011385 | 254.32 g/mol | K2O7S2 | 1 KG

Supplier: Medchemexpress LLC HYW0199701KG
Potassium pyrosulfate is an inorganic salt (pyrosulfuric acid, dipotassium salt) used as a reagent in laboratory research. It is supplied as a white to off-white solid and is intended for research use only.
- White to off-white solid
- Chemical formula K2O7S2
- Molecular weight 254.32 g/mol
- CAS number 7790-62-7
- Supplied in multiple package sizes including 1 kg
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.