Learn More
Medchemexpress LLC Potassium sodium tartrate tetrahydrate | 6381-59-5 | MFCD00150989 | 99.1% | 282.22 | 25g
SDP
Supplier:  Medchemexpress LLC HYY133225G
Encompass_Preferred
Rochelle salt is an organic-inorganic compound commonly used as a catalyst and complexing reagent It can be used to catalyze or promote certain chemical reactions and is widely used in batteries pharmaceuticals and food industries In addition this compound is also used as a substrate or intermediate in some biochemical reactions
Catalog No. 50-002-67198