Learn More
Selleck Chemical LLC Potassium sorbate S3142-100mg
SDP
Supplier:  Selleck Chemical LLC S3142100MG
Encompass_Preferred
Potassium sorbate (Sorbic acid potassium) is a nonpoisonous food preservative isolated from Sorbus aucuparia Potassium sorbate is an effective inhibitor of most molds and yeasts and some bacteria
Catalog No. 50-311-1294