Learn More
Medchemexpress LLC Potassium sulfate | 7778-80-5 | 99.2% | 174.26 | 1 KG
SDP
Supplier:  Medchemexpress LLC 1KGHYY1083H
Encompass_Preferred
Potassium sulfate can be utilized in biochemical tests for serum proteins, for determining blood potassium and other potassium salts, and for preparing protein capillary electrophoresis buffers. It is a biological material or organic compound widely used in life science research. For research use only; not sold to patients.
  • Used in biochemical tests for serum proteins.
  • Used for the determination of blood potassium and other potassium salts.
  • Used in the preparation of protein capillary electrophoresis buffers.
  • Widely used in life science research.
Catalog No. 50-003-65384