Learn More
Strem, An Ascensus Company CAS# 1312-73-8. 5g. Potassium sulfide, anhydrous, min. 95%. MFCD00043083
SDP
Supplier:  Strem, An Ascensus Company 1939005G
Encompass
CAS# 1312-73-8. 5g. Potassium sulfide, anhydrous, min. 95%. MFCD00043083. Molecular Weight: 110.26. Molecular Formula: K2S. Color/form: white to yellow pwdr. Strem# 19-3900. http://www.strem.com/catalog/v/19-3900/
Catalog No. 50-901-10650