Learn More
Research Products International Corp Potassium Tellurite, 25 Grams, Research or Further Manufacturing Use Only, Not for Food or Drug Use

Supplier: Research Products International Corp P9822025.0
- CAS Number: 7790-58-1
- Molecular Formula: K2O3Te
- Molecular Weight: 253.79
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.