Learn More
Alkali Scientific Potassium Tellurite, 25 Grams
SDP
Supplier:  Alkali Scientific ASS003CP
Encompass_Preferred
Potassium Tellurite is an inorganic salt with antimicrobial properties, commonly used in microbiology for the selective isolation of certain bacterial species. It inhibits the growth of many common bacteria, making it particularly useful for isolating and identifying pathogens in mixed microbial samples. This reagent is widely used in clinical and environmental microbiology, particularly for the isolation of pathogenic bacteria in food, water, and clinical samples. The 25-gram quantity is ideal for small-scale use in laboratories performing bacterial isolation, culture, and identification assays. Its strong antimicrobial properties make it valuable for selective enrichment procedures and pathogen detection.
Catalog No. 50-006-60753