Learn More
Medchemexpress LLC Potassium tert-butyl malonate | 75486-33-8 | 95.0% | 198.26 | 5 G

Supplier: Medchemexpress LLC HY429045G
Potassium tert-butyl malonate is an intermediate used in the synthesis of malonate monoester potassium salt. It is intended for research use only.
- Intermediate in the synthesis of malonate monoester potassium salt
- White to off-white solid appearance
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.