Learn More
Strem, An Ascensus Company CAS# 12045-78-2. 500g. Potassium tetraborate tetrahydrate, 99+%. MFCD00149924
SDP
Supplier:  Strem, An Ascensus Company 931950500G
Encompass
CAS# 12045-78-2. 500g. Potassium tetraborate tetrahydrate, 99+%. MFCD00149924. Molecular Weight: 233.44 (305.51). Molecular Formula: K2B4O7∙4H2O. Color/form: white pwdr. Strem# 93-1950. http://www.strem.com/catalog/v/93-1950/
Catalog No. 50-901-14299