Learn More
Strem, An Ascensus Company CAS# 10025-99-7. 1g. Potassium tetrachloroplatinate(II) (99.9%-Pt). MFCD00011378
SDP
Supplier:  Strem, An Ascensus Company 7819701G
Encompass
CAS# 10025-99-7. 1g. Potassium tetrachloroplatinate(II) (99.9%-Pt). MFCD00011378. Molecular Weight: 415.11. Molecular Formula: K2PtCl4. Color/form: pink to red pwdr. Strem# 78-1970. http://www.strem.com/catalog/v/78-1970/
Catalog No. 50-901-13311