Learn More
Alkali Scientific Potato Dextrose Agar (PDA) Slant 10mL Tube with Screw Cap, 10/Pk
SDP
Supplier:  Alkali Scientific PDAT01
Encompass_Preferred
Potato Dextrose Agar (PDA) Slant is a classic medium used for the cultivation and maintenance of yeasts and molds in microbiology laboratories. The nutrient base consists of potato infusion, which encourages fungal growth and promotes spore formation and pigment development important for species identification. Dextrose provides a carbohydrate energy source that supports vigorous fungal metabolism and colony development. This product is prepared as a 10 mL slant in a screw cap tube, providing a stable environment for culture storage and routine laboratory use. PDA slants are commonly used for maintaining fungal stock cultures, transferring isolates, and supporting incubation periods that may extend for several days while preventing the medium from drying out.
Catalog No. 50-006-62907