Learn More
Bachem H-Ala-Glu-OH
SDP
Supplier:  Bachem 4002318.0005
Encompass_Preferred
H-Ala-Glu-OH, 13187-90-1, C8H14N2O5, Mr 218.21, 5g. Potential degradation product of Ala-Gln, a glutamine source used in parenteral nutrition.
Catalog No. 50-194-6822