Learn More
Bachem H-Ala-D-Gln-OH
SDP
Supplier:  Bachem 4011868.0250
Encompass_Preferred
H-Ala-D-Gln-OH, 281660-34-2, C8H15N3O4, Mr 217.23, 250mg. Potential impurity of L-alanyl-L-glutamine used for glutamine supplementation.
Catalog No. 50-194-6821