Learn More
Bellco Glass, Inc. The Potter-Elvehjem tissue homogenizer has a ground glass pestle and tube. They are suitable for processing tougher tissues such as connective tissue. Clearance between the pestle and tube is .004-.006. The Potter-Elvehjem ptfe pestle is suitable for soft
SDP
Supplier:  Bellco Glass, Inc. 198320021
Encompass_Preferred
The Potter-Elvehjem tissue homogenizer has a ground glass pestle and tube. They are suitable for processing tougher tissues such as connective tissue. Clearance between the pestle and tube is .004-.006. The Potter-Elvehjem ptfe pestle is suitable for soft
Catalog No. 50-194-5200