Learn More
Aviva Systems Biology Corp POU6F1 Antibody - C-terminal region (ARP47562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POU6F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human POU6F1. Alternate names: BRN5, MPOU, TCFB1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59589