Learn More
Medchemexpress LLC Povorcitinib | 1637677-22-5 | 99.0% | C23H22F5N7O | 10MG
SDP
Supplier:  Medchemexpress LLC HY14558810MG
Encompass_Preferred
Povorcitinib is a selective JAK1 inhibitor supplied as a white to off-white solid for research use. It is used in in vitro and preclinical studies to investigate JAK/STAT signaling and inflammatory disease models. The compound has molecular formula C23H22F5N7O and a molecular weight of 507.46 g/mol.
  • Selective JAK1 inhibition for pathway-specific studies.
  • High purity (99.0%) suitable for analytical and biological assays.
  • White to off-white solid convenient for storage and formulation.
  • Available in small pack sizes for pilot and preclinical work.
  • Characterized by supporting analytical data (COA, HNMR, LCMS).
Catalog No. 50-002-58196