Learn More
Biogenex Laboratories Power Block
SDP
Supplier:  Biogenex Laboratories HK08350K
Encompass_Preferred
Power Block ™ reagent is a highly effective blocking reagent capable of reducing non-specific background staining associated with immunodetection techniques such as immunocytochemistry, ELISA, and Western blotting. Antibodies tend to bind non-specifically to membranes and tissues. To minimize this nonspecific binding, a blocking buffer is used to enhance signal-to noise ratio. Power Block™ reagent shows no cross-reactivity and it blocks more effectively than serum albumins. The blocking potency of Power Block™ reagent is superior to most other conventional formulations in producing a high signal-to-noise ratio. Power Block™ reagent may also be used as effective diluents for antibodies.""""
Catalog No. NC1244810