Learn More
Grainger Power Correction Capacitor 0.5 kvar
Supplier:  Grainger 16Y142
Encompass_Preferred
The WEG Power Factor Correction Capacitor 16Y142 is a single-phase capacitor designed to improve power factor in electrical systems. This capacitor has a capacitance of 142 microfarads and is rated for 480 volts AC. It features a rugged, impact-resistant aluminum can construction and uses non-PCB oil. The operating temperature range is -40°F to +165°F. This capacitor helps reduce energy waste and prevent overheating and voltage drops. Its compact design allows easy installation. This product helps improve power quality in commercial and industrial applications.
Catalog No. 50-276-5198