Learn More
Grainger Power Correction Capacitor 0.5 kvar
Supplier:  Grainger 16Y186
Encompass_Preferred
The Grainger Approved Power Factor Correction Capacitor is a single-phase capacitor designed to improve power factor in inductive circuits. It has a voltage rating of 240VAC and a capacitance of 30 microfarads. This capacitor connects in parallel with the load and helps offset inductive reactance to minimize wasted energy. It features a self-healing metallized film construction and quick 1/4 turn mount terminal block for easy installation. With dimensions of 2-1/2"H x 2-1/2"W x 3-3/4"D, this capacitor provides an economical solution for power factor correction in commercial and industrial applications.
Catalog No. 50-276-6224